Would you like to log in again?
VWR THERMOMETER BLK T-SHAPE
Supplier: VWR International
VWR Economy Digital Thermometer^ Temperature Range Is -50 To 150C/-58 To 302F, Resolution 0.1. Stainless Steel Probe Length 5 Inches Supplied With Protective Sleeve. On/Off And Hold Function. C/F Selectable.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR THERMOMETER BLK T-SHAPE 89140-526 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty