Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Economy Timer
Supplier: VWR
VWR®
98771-09
89140-544EA 31.42 USD InStock
89140-544
VWR® Economy Timer
Timers Digital Timers
Timer can be programmed in countdown mode or countup mode up to 99 minutes, 59 seconds.
  • Count up and down simultaneously
  • Big digit display
Delivery information: Supplied with loud buzzer, magnet, clip, and stand.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty