Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Four-Channel Economy Timer
Supplier: Avantor
VWR®
98771-10
89140-552EA 41.33 USD InStock
89140-552
VWR® Four-Channel Economy Timer
Timers Digital Timers
Time-of-day clock.
  • Loud alarm
  • Memory function remembers the last time set
  • Fold-out stand and magnet included
Counts up or down simultaneously for 99 hours, 59 minutes, 59 seconds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty