Would you like to log in again?
VWR HYGRO/THERMO PNL MNT ECON
Supplier: VWR International
VWR Economy Hygrometer/Thermometer^ Temperature Probe Range -50 To 70C/-58 To 158F, . Resolution 0.1. Humidity Range 20 To 99%, Resolution 1%. Min/Max Memories. C/F Selectable. Ice Point Alert.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR HYGRO/THERMO PNL MNT ECON 89140-556 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty