Would you like to log in again?
About Face™ Cleansing Wipes, Micronova
Supplier: Micronova
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
About Face™
MMR-01
89140-690CS 406.35 USD InStock
89140-690
About Face™ Cleansing Wipes, Micronova
Wipes Disposable Wipes and Towels Facial Tissues
A pre-moistened cleansing wipe that is gentle enough for the face and that can be used to remove surface soil prior to gloving or sanitizing hands.
  • Individually packaged, polyblend cleansing wipe
  • Pre-moistened with a gentle, alcohol-free cleansing lotion
  • Highly effective as a make-up remover
  • Removes surface soil prior to gloving or sanitizing hands
About Face™ Cleansing Wipes are individually packaged and ideal for pre-gowning rooms.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty