Would you like to log in again?
M-Zone MegaSorb™ Microfiber String Mops, Micronova
Supplier: Micronova
MegaSorb™
MZCRMS-10 MZCRMS11-12 JZMZCRMS-10
10030-340CS 412.57 USD InStock
10030-340 89092-124 89140-706
M-Zone MegaSorb™ Microfiber String Mops, Micronova
Mops
A highly absorbent mop that combines the cleaning efficiency of microfiber with the ease of a looped string mop. The tufted strands are comprised of polyester and nylon filaments, designed to pick up liquids and tiny particles in controlled and critical environments.
  • Highly absorbent and low-linting
  • Polyester and nylon tufted microfiber tubes
  • Picks up tiny particles where cotton fibers just roll over
  • White microfiber option includes an additional scrubbing band
  • Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
The M-Zone™ MegaSorb™ Microfiber String Mop features distinctive microfiber tubes with tufted strands made of polyester and nylon filaments, 30 times smaller than cotton fibers, that work to pick up tiny particles.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty