Would you like to log in again?
VWR® Self-Seal Sterilization Pouches
Supplier: VWR
VWR®
89140-800 89140-804 89140-802
89140-800CS 623.76 USD InStock
89140-800 89140-804 89140-802
VWR® Self-Seal Sterilization Pouches
Bags Sterilization Pouches
Sterilization pouches are made of surgical-grade paper and blue polypropylene/polyester film.
  • Color-coded indicators on each pouch
  • Sterilizable by steam or ETO
Easy-to-open lip closure, prefold self-adhesive strip, and flat seals ensure seal integrity.
Certifications: Indicator ink conforms to ISO 11140-1. Manufactured in a facility with ISO 9001:2008, ISO 13485, and HACCP certifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty