Would you like to log in again?
Viral Nucleic Acid Extraction Kits, IBI Scientific
Supplier: IBI Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IB47403 IB47402 IB47401 IB47400
89140-844EA 301.41 USD InStock
89140-844 89140-842 89140-848 89140-846
Viral Nucleic Acid Extraction Kits, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
IBI's Viral Nucleic Acid Extraction Kit was designed specifically for purification of viral DNA/RNA from cell-free samples; such as serum, plasma, body fluids, and the supernatant of viral infected cell cultures
  • Sample Size: 200 µl sample (plasma, serum, body fluid, or supernatant of viral infected cell cultures)
  • Operation Time: 40 min or less
  • Elution Volume: 50 µl
The Viral Nucleic Acid Extraction Kit was designed specifically for purification of viral DNA/RNA from cell-free samples such as: serum, plasma, body fluids, and the supernatant of viral infected cell cultures. DNA/RNA viruses are lysed quickly and efficiently using the lysis buffer, which is a highly concentrated solution of chaotropic salt. When combined with ethanol, the AD Buffer creates an optimum condition for nucleic acid binding to the glass fiber of the column. Contaminants such as salts, metabolites, and soluble macromolecular cellular components are removed in the wash process. The nucleic acids are eluted in RNase-Free water and are then ready for use in subsequent reactions including Real-Time PCR, Automated Fluorescent DNA Sequencing, PCR, and other enzymatic reactions. The detection limit for certain viruses depends on the sensitivity of the individual PCR or RT-PCR assays.
Certifications: For research use only.
Caution: Components contain irritant agents. Always wear a lab coat, disposable gloves, and protective goggles during use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty