Would you like to log in again?
TXB2 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-002
89141-054EA 503 USD InStock
89141-054
TXB2 ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of TXB2 in culture supernatants, plasma, serum, saliva, and urine from any species
The TXB2 ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 in a convenient 96 well plate based assay. This kit is highly cited in peer-reviewed publications in a diverse assortment of sample types including mucosa, tissue, serum, plasma, urine, and much more. The TXB2 ELISA kit provides easy-to-use stable liquid color-coded reagents that reduce errors.
Cited sample types include cell lysate, coronary effluent, liver perfusate, platelets, rectal dialysate, and whole blood.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-270 (5x96 wells)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty