Would you like to log in again?
6-keto-PGF1α ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-004
89141-056EA 489 USD InStock
89141-056
6-keto-PGF1α ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ELISA, detecting as low as 1.40pg/ml in just 3 hours.
For the quantitative determination of 6-keto-PGF1α in culture supernatants, serum, saliva, and urine from any species.
The 6-keto-PGF1α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 1α EIA kit has been fully validated in-house in a wide variety of sample matrices.
Ordering information: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-272 (5x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty