Would you like to log in again?
8-iso-PGF2α ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-010
89141-062EA 517 USD InStock
89141-062
8-iso-PGF2α ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of 8-iso-PGF2α in culture supernatants, tissue, and urine from any species
The measurement of F2-isoprostanes has emerged as one of the most reliable approaches to assess oxidative stress status in vivo, providing an important tool to explore the role of oxidative stress in the pathogenesis of human disease. This is a colorimetric competitive immunoassay kit for the quantitative determination of free 8-iso-Prostaglandin F2α in biological fluids such as urine and in culture supernatants. To measure 8-iso-Prostaglandin F2α levels in serum, plasma or other tissue samples, use the Direct 8-iso-PGF2α EIA Kit. 8-iso-PGF2α is produced in vivo by both non-cyclooxygenase and cyclooxygenase dependant mechanisms from arachidonic acid. It has been shown to be a potent vasoconstrictor, a potential mediator of hepatorenal syndrome and a mutagen in 3T3 cells and vascular smooth muscle cells. 8-iso-PGF2α has been shown to circulate in plasma and is excreted in urine.
Cited sample type includes plasma.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-278 (5x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty