Would you like to log in again?
Endothelin-1 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ADI-900-020A
89141-078EA 756 USD InStock
89141-078
Endothelin-1 ELISA Kit, Enzo Life Sciences
Assays ELISAs
The Endothelin-1 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours
  • Highly sensitive measurement
  • Higher throughput format
  • Flexible option of overnight or 1 hour primary incubation
  • Suitable for a variety of sample matrices
For the quantitative determination of ET-1 in culture supernatants, plasma, and serum of human, mouse, rat, cow, dog, pig, and rabbit origin.
This ultra-sensitive kit lets you quantify as little as 0.41 pg/mL ET-1 in a variety of biological matrices with the option of overnight or a 1 hour primary incubation.
Delivery information: Kit contains microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty