Would you like to log in again?
IL-10 (Human), ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-036 ADI-901-036
89141-298EA 2757.89 USD InStock
89141-298 89141-092
IL-10 (Human), ELISA kit, Enzo Life Sciences
Assays ELISAs
The IL-10 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours
For the quantitative determination of human IL-10 in culture supernatants, plasma, and serum.
Absorbance is read at 450 nm. Ready-to-use liquid color-coded reagents reduce laboratory errors.
Delivery information: Kit includes microtiter plate, conjugate, antibody, assay buffer 13 concentrate, wash buffer concentrate, standard, TMB substrate, stop solution 2
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty