Would you like to log in again?
IL-6 (mouse), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-045
89141-098EA 634 USD InStock
89141-098
IL-6 (mouse), ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of mouse Il-6 in culture supernatants and serum
The IL-6 (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours.
Cited sample type includes tissue.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty