Would you like to log in again?
TNF Alpha (Mouse) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-047
89141-100EA 669 USD InStock
89141-100
TNF Alpha (Mouse) ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of mouse TNF-α in culture supernatants and serum
The TNF-α (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 5 hours. Absorbance is read at 450nm.
Cited sample type includes tissue and plasma.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 16, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty