Would you like to log in again?
15(S)-HETE ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-051
89141-104EA 499 USD InStock
89141-104
15(S)-HETE ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of 15(S)-HETE in culture supernatants, plasma, serum, and urine from any species.
The 15(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 405nm.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-302 (5x96 wells)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty