Would you like to log in again?
Cysteinyl leukotriene ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-070
89141-122EA 532 USD InStock
89141-122
Cysteinyl leukotriene ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of Cysteinyl Leukotrienes (LTC4/D4/E4) in culture supernatants and urine from any species.
The Cysteinyl Leukotriene EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405nm. Safe non-radioactive measurement of LTC4/D4/E4. Ready-to-use liquid color-coded reagents reduce errors.
Ordering information: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-318 (5x96 wells)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty