Would you like to log in again?
Cortisol ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-071 ADI-900-071
89141-320EA 1850 USD InStock
89141-320 89141-124
Cortisol ELISA Kit, Enzo Life Sciences
Assays ELISAs
Widely used sensitive cortisol ELISA kit with a large variety of sample types and highly reproducible results for stress, autoimmune disease and cancer research.
For the quantitative determination of Cortisol in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample type includes feces.
The Cortisol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit is highly cited in peer-reviewed publications. The ready-to-use liquid color-coded reagents provided are safe, non-radioactive, and reduce error in the lab. The broad dynamic range lets you accurately measure Cortisol levels in a variety of sample matrices.
Ordering information: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-320 (5x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty