Would you like to log in again?
Big Endothelin-1 (Rat), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-073
89141-126EA 1023.55 USD InStock
89141-126
Big Endothelin-1 (Rat), ELISA Kit, Enzo Life Sciences
Assays ELISAs
The Big Endothelin-1 (Rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results overnight + 1 hour.
  • Measure low levels of rat big endothelin-1
  • Convenient method
  • Assay up to 38 samples in duplicate per plate
For the quantitative determination of rat Big ET-1 in culture supernatants, plasma, and serum.
Delivery information: Kit contains microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty