Would you like to log in again?
COX-2 (Human), ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ADI-900-094
89141-144EA 1080 USD InStock
89141-144
COX-2 (Human), ELISA kit, Enzo Life Sciences
Assays ELISAs
The COX-2 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours
  • Easy to use
  • lThe color-coded reagents reduce errors
  • Reliable and fast
For the quantitative determination of human COX-2 in cell lysates.
This is the most sensitive COX-II immunoassay kit commercially available for inflammation research.
Delivery information: Supplied with microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty