Would you like to log in again?
Corticosterone ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-097 ADI-900-097
89141-146EA 445.66 USD InStock
89141-146 101106-156
Corticosterone ELISA kit, Enzo Life Sciences
Assays ELISAs
The Corticosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours
  • Sensitive measurement of Corticosterone
  • High throughput results
  • Routinely used to monitor stress levels in zoo animals
This is widely used sensitive corticosterone ELISA kit with a large variety of sample types and highly reproducible results for animal stress research. For the quantitative determination of Corticosterone in culture supernatants, feces, plasma, serum, and saliva from any species. Cited sample types include cell lysate and whole blood.
Absorbance is read at 405 nm. The Corticosterone EIA kit has low cross-reactivity with related steroids. The broad dynamic range makes this kit ideal for a wide variety of sample matrices from any species
Ordering information: Alternative size available as ADI-900-097 (1x96 wells)
Delivery information: Kit includes DxS microtiter plate, conjugate, antibody, assay buffer 15 concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution, steroid displacement reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty