Would you like to log in again?
Estriol ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-100
89141-152EA 379 USD InStock
89141-152
Estriol ELISA Kit, Enzo Life Sciences
Assays ELISAs
Certified for a large variety of sample types for steroid and hormone research.
For the quantitative determination of Estriol in culture supernatants, plasma, serum, saliva, and urine from any species.
The Estriol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit has a broad dynamic range suitable for a wide variety of sample matrices and the ability to <60 pg/mL estriol.
Ordering information: Contains GxR IgG Microtiter plate, Assay buffer concentrate, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty