Would you like to log in again?
IgG2b (mouse), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-110
89141-162EA 532 USD InStock
89141-162
IgG2b (mouse), ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of IgG2b in culture supernatants and serum of mouse origin.
The IgG2b (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minutes. Ideal for determining the isotype and concentration of monoclonal antibodies.
Ordering information: Contains GxM IgG Microtiter plate, mouse IgG2b conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgG2b standard, TMB Substrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty