Would you like to log in again?
IFN-γ (Human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-136
89141-194EA 792 USD InStock
89141-194
IFN-γ (Human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
The IFN-gamma (Human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.
  • Ultrasensitive
  • Efficient - only 50 µL diluted sample needed per well
  • Measure natural and recombinant human IFN-γ
For the quantitative determination of human IFN-gamma in culture supernatants, plasma, serum, and urine.
Absorbance is read at 450 nm.
Delivery information: Supplied with microtiter plate, SA-HRP concentrate, SA-HRP dilution buffer, biotinylated antibody, standard, standard diluent, wash buffer concentrate, TMB substrate, stop solution
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty