Would you like to log in again?
[pSer15]HSP27 (human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-170
89141-248EA 735 USD InStock
89141-248
[pSer15]HSP27 (human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
Only commercially available ELISA kit for ultra-sensitive detection of human phospho-Ser15 HSP27 in cell lysates, plasma, and serum.
For the quantitative determination of human [pSer15]HSP27 in cell lysates, plasma, and serum.
The [pSer15]HSP27 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 27, Wash buffer concentrate, Standard, TMB Substrate, 5X Extraction reagent, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty