Would you like to log in again?
17β-Estradiol ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-008
89141-276EA 1500 USD InStock
89141-276
17β-Estradiol ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ELISA, measuring as little as 28.5 pg/mL of 17β-Estradiol in < 3 hours.
For the quantitative determination of 17β-Estradiol in culture supernatants and saliva from any species. For serum and plasma use 89141-346.
The 17β-Estradiol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. This kit offers specific and save non-radioactive quantification of estradiol with ready-to-use liquid color-coded reagents to reduce error.
Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-060 (1x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty