Would you like to log in again?
15-deoxy-δ12,14-PGJ2 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-023
89141-290EA 2350 USD InStock
89141-290
15-deoxy-δ12,14-PGJ2 ELISA Kit, Enzo Life Sciences
Assays ELISAs
First-to-market 15-d-PGJ2 ELISA commercially available for eicosanoid research.
For the quantitative determination of 15-deoxy-Δ12,14-PGJ2 in culture supernatants, plasma, saliva, and urine from any species.
The 15-deoxy-Δ12,14-PGJ2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit saves time and is much less time consuming than GC/MS analysis. Ready-to-use liquid color-coded reagents reduce laboratory error.
Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-082 (1x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty