Would you like to log in again?
LTB4 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-068
89141-314EA 2000 USD InStock
89141-314
LTB4 ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of LTB4 in culture supernatants, plasma, saliva, and urine from any species
The LTB4 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Assay up to 39 samples in duplicate with a broad dynamic range that is suitable for a wide variety of sample matrices. Document your results to scientists world-wide with this highly cited kit.
Cited sample types include bronchioalveolar lavage fluid, cell lysate, crevicular fluid, tissue, serum and whole blood.
Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-118 (1x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty