Would you like to log in again?
PGF2α ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-069
89141-316EA 1940 USD InStock
89141-316
PGF2α ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of PGF2α in culture supernatants, plasma, serum, saliva, and urine from any species
The PGF2α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Broad dynamic range is suitable for a wide variety of sample types. Ready-to-use liquid color-coded reagents are safe, non-radioactive, and reduce laboratory errors.
Cited sample types include cell lysate and whole blood.
Ordering information: Contains DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-120 (1x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty