Would you like to log in again?
DHEA ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-093
89141-326EA 1974.48 USD InStock
89141-326
DHEA ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of DHEA in culture supernatants, plasma, serum, saliva, and urine from any species.
The DHEA EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit has low reactivity to structurally related molecules and offers sensitive measurement of DHEA in a variety of sample types.
Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-142 (1x96 wells).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty