Would you like to log in again?
TNF Alpha (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-099 ADI-901-099
89141-330EA 2751.28 USD InStock
89141-330 89141-150
TNF Alpha (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ELISA for measuring TNF-α, the primary mediator of immune regulation.
For the quantitative determination of human TNF-α in culture supernatants, plasma, and serum.
The TNF-α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450 nm. Assay up to 39 samples in duplicate per 96-well plate. The broad dynamic range makes assaying different pathologies easy.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 21, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty