Would you like to log in again?
Aldosterone ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-173 ADI-900-173
89141-254EA 440 USD InStock
89141-254 89141-344
Aldosterone ELISA kit, Enzo Life Sciences
Assays ELISAs
The Aldosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.
  • Highly sensitive
  • Negligible reactivity with other steroids
  • Fully quantitative results that surpass semi-quantitative Western blots
For the quantitative determination of Aldosterone in plasma, serum, and urine from any species.
Delivery information: Kit includes DxS microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty