Would you like to log in again?
17β-Estradiol High Sensitivity ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-174 ADI-900-174
89141-256EA 434 USD InStock
89141-256 89141-346
17β-Estradiol High Sensitivity ELISA Kit, Enzo Life Sciences
Assays ELISAs
The 17beta-Estradiol (Serum/Plasma) EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours.
  • Highly sensitive measurement
  • Fast and reliable results
  • ully quantitative results that surpass semi-quantitative Western blot analysis
  • Convenient pre coated plate and color coded reagents reduces errors
For the quantitative determination of 17beta-Estradiol in plasma and serum from any species.
Delivery information: Kit includes DxS microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty