Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APMAP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-714-100
89141-368EA 528 USD InStock
89141-368
Anti-APMAP Rabbit Polyclonal Antibody
Antibodies
Adipose tissue is the source for a variety of bioactive molecules that regulate metabolism and immunity termed adipokines, most of which are elevated in the circulation of obese individuals. Adiponectin functions as an insulin sensitizing factor, resulting in reduced hepatic glucose production, increased glucose uptake in the muscle, and increased fatty acid oxidation in the liver and muscle. In contrast to most adipokines, adiponectin is detected at high levels in normal human plasma, but is often reduced in obese subjects and in those with increased insulin resistance and type 2 diabetes. Due to its suggested protective role in insulin resistance, adiponectin may be a useful pharmacological target for various metabolic diseases.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat Adiponectin. Detects a band of ~32kDa by Western blot.

Type: Primary
Antigen: APMAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty