Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BECN1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-721-100
89141-382EA 525 USD InStock
89141-382
Anti-BECN1 Rabbit Polyclonal Antibody
Antibodies
Beclin-1 is a coiled-coil Bcl-2-interacting protein that functions in autophagy, the process of bulk degradation of cellular proteins. Beclin-1 is homologous to the yeast autophagy gene apg6, and is expressed at reduced levels in human breast carcinoma, suggesting that defects in autophagy proteins may contribute to the development or progression of tumors. Bcl-2 can inhibit Beclin-1-dependent autophagy in yeast and mammalian cells, suggesting that Bcl-2 functions as both an anti-autophagic and anti-apoptotic protein.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat Beclin-1. Detects a band of ~57kDa by Western blot.

Type: Primary
Antigen: BECN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty