Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAG1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-735-100
89141-410EA 514 USD InStock
89141-410
Anti-BAG1 Rabbit Polyclonal Antibody
Antibodies
Bag-1 is a multifunctional Bcl-2-binding protein that lacks sequence homology with Bcl-2 and its related proteins, and functions to enhance the anti-apoptotic effects of Bcl-2. Bag-1 binds to the intracellular domain of the hepatocyte growth factor (HGF) and platelet-derived growth factor (PDGF) receptors, enhancing growth factor mediated protection from apoptosis. Bag-1 is known to modulate the chaperone activity of Hsc70/Hsp70, serving as a nucleotide exchange factor via binding to the ATPase domain of Hsc70/Hsp70. Bag-1 is known to promote cell survival through activation of the Raf-1 kinase.

  • For Western Blot

Recognizes human, mouse and rat Bag-1. Detects a band of ~37kDa by Western blot.

Type: Primary
Antigen: BAG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty