Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GSK3A Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-759-100
89141-420EA 556 USD InStock
89141-420
Anti-GSK3A Mouse Monoclonal Antibody
Antibodies
Glycogen Synthase Kinase 3 (GSK3) exists in two isoforms, alpha and beta (GSK3α, and GSK3β). GSK3 is an active kinase that becomes phosphorylated by PKB/akt upon growth factor stimulation. Phosphorylation of GSK3α at serine 21 or GSK3β at serine 9, respectively, leads to inhibition of kinase activity of the GSK3.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse, rat and dog GSK-3alpha.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: GSK-3ALPHA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty