Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Endothelin B Receptor Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-791-100
89141-446EA 442 USD InStock
89141-446
Anti-Endothelin B Receptor Rabbit Polyclonal Antibody
Antibodies
Endothelins are vasoactive peptides that exist in three forms (ET-1, ET-2, and ET-3), and function in the maintenance of vascular tone via two identified seven transmembrane-spanning domain-containing G protein-coupled receptors (GPCR), endothelin A (ETA) and endothelin B (ETB). The human ETA receptor is primarily expressed in vascular smooth muscle cells, including those of the heart (also in myocytes), lung, and brain, and its stimulation generally results in vasoconstriction. Activation of ETA receptors can be stimulated by binding of ET-1 and ET-2, but not ET-3. triggering the release of vasorelaxive factors such as nitric oxide (NO) and prostanoids from endothelial cells. Human ETB receptors are primarily expressed in endothelial cells lining the vessel walls of the lungs, heart, and brain, and in contrast to ETA receptors, stimulation generally results in transient vasodilation. Activation of ETB receptors can be stimulated by binding of all three endothelins, triggering the release of vasorelaxive

  • For Immunohistochemistry, Western Blot

Recognizes rat endothelin receptor ETB. Predicted MW of ~50kDa. Higher MW species (~ 55kDa) and intermediates also observed by Western blot which reflect post-translational modification of the receptor. Predicted species reactivity based on epitope sequence homology: human, mouse, prairie deer mouse, beach mouse, rhesus macaque, chimpanzee, horse, dog.

Ordering information: Manufactured by Proteimax.

Type: Primary
Antigen: ENDOTHELIN B RECEPTOR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty