Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BLTR2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-794-100
89141-452EA 406 USD InStock
89141-452
Anti-BLTR2 Rabbit Polyclonal Antibody
Antibodies
Leukotrienes are metabolic products of arachidonic acid formed by the action of 5-lipoxygenase (5-LO), with the principal products being leukotrienes C4 (LTC4) and B4 (LTB4). LTB4 is a dihydroxy-leukotriene produced in a variety of cells including eosinophils, neutrophils, platelets, and vascular cells, and is a potent leukocyte chemoattractant known to regulate leukocyte activation, cytokine secretion, inflammatory gene transcription, and IgE synthesis. As such, LTB4 has been implicated in a number of disease pathologies including atherosclerosis, asthma, psoriasis, and inflammatory bowel disease. The many actions of LTB4 are associated with two identified mammalian receptors, BLT1 and BLT2. BLT1 is a high-affinity LTB4 receptor expressed on peripheral leukocytes, while BLT2 is a more ubiquitously expressed low affinity LTB4 receptor whose function is not well understood. The BLT1 receptor is transiently expressed by antigen-primed T-cells in humans, and its expression is known to play a critical role in collagen-induced arthritis in mice.

  • For Western Blot

Recognizes rat LTB4 Receptor (BLT2). Predicted MW of ~38kDa. Higher MW species (~50kDa) and intermediates also observed by Western blot which reflect post-translational modification of the receptor. Predicted species reactivity based on epitope sequence homology: human, mouse, horse, chimpanzee, rhesus macaque, wild boar, dog, platypus.

Ordering information: Manufactured by Proteimax.

Type: Primary
Antigen: BLTR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty