Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Neuropeptide Y Receptor Type 1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-798-100
89141-460EA 422 USD InStock
89141-460
Anti-Neuropeptide Y Receptor Type 1 Rabbit Polyclonal Antibody
Antibodies
Neuropeptide Y (NPY) and the closely related peptide YY (PYY) and pancreatic polypeptide (PP) are a family of 36-amino acid, tyrosine-rich peptides with amidated C-terminal ends that are known to regulate gastrointestinal and central nervous system functions including food intake and the inhibition of anxiety. Four functional NPY receptors (Y1/Y2/Y4/Y5) have been identified to date, with a fifth (y6) being a non-functional receptor in primates that is absent from the rat genome. Y1, Y2, and Y5 receptors preferentially bind NPY and PYY, while the Y4 receptor preferentially binds PP. All four receptors signal via coupling to Gi/o alpha subunits, inhibiting adenylyl cyclase activity, thereby decreasing intracellular cAMP levels. Y1 receptor deficient mice display a variety of physical and behavioral phenotypes including obesity, hyperinsulinemia, and increased territorial behavior.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse and rat neuropeptide Y receptor Y1. Predicted MW of ~44kDa. Higher MW species (~80kDa) also observed by Western blot which may reflects post-translational modification or dimerization of the receptor. Predicted species reactivity based on epitope sequence homology: horse, chimpanzee, rhesus macaque, guinea pig.

Ordering information: Manufactured by Proteimax.

Type: Primary
Antigen: NEUROPEPTIDE Y RECEPTOR TYPE 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty