Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APP Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-803-100
89141-470EA 508 USD InStock
89141-470
Anti-APP Mouse Monoclonal Antibody
Antibodies
The beta-amyloid peptide (beta A4), proteolytically released from the amyloid precursor protein (APP), provides the principal component of senile plaques in Alzheimer's disease. Cleavage of APP by alpha-secretase or alternatively by beta-secretase leads to generation and extracellular release of soluble APP peptides, S-APP-alpha and S-APP-beta, respectively, and the retention of corresponding membrane-anchored C-terminal fragments, C83 and C99. Subsequent processing of C83 by gamma-secretase yields P3 peptides. This is the major secretory pathway and is nonamyloidogenic. Alternatively, presenilin/ nicastrin-mediated gamma-secretase processing of C99 releases the amyloid beta proteins, amyloid-beta 40 (Abeta40) and amyloid-beta 42 (Abeta42), major components of amyloid plaques, and the cytotoxic C-terminal fragments, gamma-CTF(50), gamma-CTF(57) and gamma-CTF(59).

  • For ELISA, Flow Cytometry, Immunocytochemistry, Western Blot

Recognizes human amyloid beta A4 (CT, 40/42). Cross-reacts with APP. Detects a band of ~4kDa by Western blot.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty