Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Vasopressin Receptor 1A Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-811-100
89141-474EA 453 USD InStock
89141-474
Anti-Vasopressin Receptor 1A Rabbit Polyclonal Antibody
Antibodies
Arginine vasopressin (AVP) is a nonapeptide hormone produced in magnocellular neurons of the hypothalamus, which functions as an antidiuretic in the kidney and systemically as a regulator of vasoconstriction, blood volume, and blood pressure. AVP acts by binding to a set of specific seven-transmembrane domain-containing G-protein coupled receptors (GPCR) classified into three groups: V1a (V1) receptors regulate vasoconstriction, and are expressed in vascular smooth muscle, kidney, myometrium, bladder, adipocytes, hepatocytes, platelets and testis; V1b (V3) receptors stimulate adrenocorticotropic hormone (ACTH) release in the anterior pituitary, but may also participate in corticotropin-releasing hormone (CRH) release by the hypothalamus; and V2 receptors of the kidney regulate the antidiuretic effects of AVP. The V1b receptor signals through coupling of Gq/11 alpha subunits, activating phospholipase C and release of intracellular Ca2+ via phosphoinositide second messengers. V1b receptor overexpression is a common marker for ACTH-secreting corticotropic adenomas.

  • For Western Blot

Recognizes human, mouse and rat vasopressin receptor V1a. Predicted MW of ~48kDa. Higher MW species (~50kDa) also observed by Western blot which reflects post-translational modification of the receptor. Predicted species reactivity based on epitope sequence homology: rhesus macaque, dog, bovine.

Ordering information: Manufactured by Proteimax.

Type: Primary
Antigen: VASOPRESSIN RECEPTOR 1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty