Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PGF2 alpha Receptor Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-901-100
89141-654EA 481 USD InStock
89141-654
Anti-PGF2 alpha Receptor Rabbit Polyclonal Antibody
Antibodies
Arachidonic acid is converted by cyclooxygenase (COX) to prostaglandin H2 (PGH2), which is further converted to the derivative prostanoids PGD2, PGE2, PGF2α, PGI2 (prostacyclin) and thromboxane A2 (TXA2). Five classes of prostanoid receptors are recognized and named DP, EP, FP, IP and TP receptors corresponding to the agonist properties of the associated prostanoids. FP receptors are found in smooth muscle where they mediate contraction, and are believed to be coupled via Gq/11 subunits to stimulate phosphatidyl inositol (PI) hydrolysis. The FP agonist Fluprostenol (a PGF2α analogue) is used in animal husbandry to synchronize oestrus and induce partruition. FP receptors are known to mediate intraocular pressure, and FP agonists such as latanoprost and travoprost are drugs commonly use to treat glaucoma.

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes mouse and rat PGF2 alpha Receptor (FP). Predicted MW of ~41kDa, observed by Western blot ~45kDa. Higher MW species (~60kDa) and intermediates also observed by Western blot which reflect post-translational modification of the receptor. Predicted species reactivity based on epitope sequence homology: wild boar, dog.

Ordering information: Manufactured by Proteimax.

Type: Primary
Antigen: PGF2 ALPHA RECEPTOR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty