Would you like to log in again?
Acridinium Ester, (Trigger Solution), Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-906-001
89141-658EA 189.44 USD InStock
89141-658
Acridinium Ester, (Trigger Solution), Enzo Life Sciences
Electrophoresis Reagents Western Blotting Reagents
Species Independent
Liquid. Solution 1 contains hydrogen peroxide in nitric acid. Solution 2 contains cetyl trimethylammonium chloride in sodium hydroxide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty