Would you like to log in again?
sPLA2 Activity Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-907-002
89141-664EA 597 USD InStock
89141-664
sPLA2 Activity Kit, Enzo Life Sciences
Assays Cellular Assays
For the quantitative determination of sPLA2 in culture supernatants, cell lysates, and buffers from any species.
The sPLA2 activity kit is a higher throughput assay for the quantification of up to 42 samples in duplicate per 96 well kit. This kit provides safe, sensitive, and reliable results in under an hour.
Ordering information: Contains Microtiter plate, Standard, Reaction buffer concentrate, Substrate, Stop reagent, Color reagent.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty