Would you like to log in again?
Hydrogen Peroxide Chemiluminescent Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-907-012
89141-668EA 452 USD InStock
89141-668
Hydrogen Peroxide Chemiluminescent Detection Kit, Enzo Life Sciences
Assays Cellular Assays
For the quantitative determination of hydrogen peroxide in culture supernatants and cell lysates from any species.
The Hydrogen Peroxide chemiluminescent detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide. This kit offers more sensitivity over the Hydrogen Peroxide colorimetric detection kit but is not recommended for samples that contain phenol red or hemoglobin.
Ordering information: Contains White Microtiter plate, Standard, Substrate, Trigger solution.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty