Would you like to log in again?
Caspase-3 Colorimetric Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-907-013
89141-670EA 605 USD InStock
89141-670
Caspase-3 Colorimetric Detection Kit, Enzo Life Sciences
Assays Cellular Assays
For the colorimetric measurement of Caspase-3 in cell lysates and buffers from any species.
The Caspase-3 colorimetric detection kit provides a higher throughput method for measuring caspase-3 activity in up 37 samples in duplicate per kit. The kit is flexible, providing both kinetic or end-point assay options.
Ordering information: Contains Half-Area Microtiter plate, Standard, DTT, p-Nitroaniline Calibrator, Reaction buffer concentrate, Substrate concentrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty