Would you like to log in again?
Hydrogen Peroxide Colorimetric Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-907-015
89141-674EA 431.92 USD InStock
89141-674
Hydrogen Peroxide Colorimetric Detection Kit, Enzo Life Sciences
Environmental Test Kits
For the quantitative determination of hydrogen peroxide in culture supernatants, serum, plasma, and urine from any species
The Hydrogen Peroxide colorimetric detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide in a wide variety of samples.
Cited sample type includes tissue.
Ordering information: Contains Half-Area Microtiter plate, Standard, Color reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty