Would you like to log in again?
Nitric Oxide (Total), Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-917-020
89141-754EA 431.43 USD InStock
89141-754
Nitric Oxide (Total), Detection Kit, Enzo Life Sciences
Assays Cellular Assays
For the quantitative determination of total Nitric Oxide in culture supernatants, plasma, serum, saliva, and urine from any species
The Nitric Oxide (total), detection kit is based on the enzymatic conversion of nitrate to nitrite by the enzyme nitrate reductase, followed by the Griess reaction to form a colored azo dye product. Quantification is performed by measuring absorption at 540-570 nm. Only 0.050 mL diluted sample needed per well saves precious sample.
Cited sample type includes tissue.
Ordering information: Contains Microtiter plate, Nitrate Reductase, Nitrate Reductase Storage Buffer, NADH, Nitrate Standard, Reaction buffer concentrate, Griess Reagent I, Griess Reagent II.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty