Would you like to log in again?
PGE2 FPIA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-920-001
89141-756EA 375 USD InStock
89141-756
PGE2 FPIA Kit, Enzo Life Sciences
Assays Immunological Assays
For the quantitative determination of PGE2 in culture supernatants from any species.
The PGE2 FPIA kit is a homogeneous mix-and-read fluorescence polarization enzyme immunoassay kit with. Results are obtained within just 30 minutes and the generated signal is stable for up to 20 hours. The PGE2 FPIA kit is ideal for high throughput screening (HTS) applications. A convenient homogeneous mix-and-read assay that is amendable to miniaturization and automation, this kit has a robust Z factor >0.8.
Ordering information: Contains Conjugate concentrate, Antibody, Assay buffer concentrate, Standard.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty